missing translation for 'onlineSavingsMsg'
Learn More
Learn More
5-Norbornen-2-yl Acetate (endo- and exo- mixture) 96.0+%, TCI America™
Supplier: TCI America N0895100G
Specifications
| 5-Norbornen-2-yl Acetate (endo- and exo- mixture) | |
| Yellow | |
| C9H12O2 | |
| 100 g | |
| DRWRVXAXXGJZIO-UHFFFAOYSA-N | |
| 5-bicyclo[2.2.1]hept-2-enyl acetate | |
| 95688 | |
| ≥96.0% (GC) |
| 6143-29-9 | |
| 73°C | |
| MFCD00167565 | |
| 5-norbornen-2-yl acetate, 5-norbornene-2-yl acetate, 2-acetoxy-5-norbornene, bicyclo 2.2.1 hept-5-en-2-yl acetate, bicyclo 2.2.1 hept-5-en-2-ol, acetate, acetic acid bicyclo 2.2.1 hept-5-en-2-yl ester, 5-norbornen-2-yl acetate, mixture of endo and exo, 5-norbonen-2-yl acetate, 5-norbornen-2-ol, acetate | |
| CC(=O)OC1CC2CC1C=C2 | |
| 152.193 | |
| 152.19 | |
| Liquid |
Chemical Identifiers
| 6143-29-9 | |
| 152.193 | |
| DRWRVXAXXGJZIO-UHFFFAOYSA-N | |
| 95688 | |
| CC(=O)OC1CC2CC1C=C2 |
| C9H12O2 | |
| MFCD00167565 | |
| 5-norbornen-2-yl acetate, 5-norbornene-2-yl acetate, 2-acetoxy-5-norbornene, bicyclo 2.2.1 hept-5-en-2-yl acetate, bicyclo 2.2.1 hept-5-en-2-ol, acetate, acetic acid bicyclo 2.2.1 hept-5-en-2-yl ester, 5-norbornen-2-yl acetate, mixture of endo and exo, 5-norbonen-2-yl acetate, 5-norbornen-2-ol, acetate | |
| 5-bicyclo[2.2.1]hept-2-enyl acetate |
Safety and Handling
TSCA : No