missing translation for 'onlineSavingsMsg'
Learn More

5-Norbornen-2-yl Acetate (endo- and exo- mixture) 96.0+%, TCI America™

Catalog No. p-7134014
Change view
Click to view available options
Quantity:
25 g
100 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
N089525G 25 g
N0895100G 100 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. N089525G Supplier TCI America Supplier No. N089525G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 6143-29-9
Molecular Formula C9H12O2
Molecular Weight (g/mol) 152.193
MDL Number MFCD00167565
InChI Key DRWRVXAXXGJZIO-UHFFFAOYSA-N
Synonym 5-norbornen-2-yl acetate, 5-norbornene-2-yl acetate, 2-acetoxy-5-norbornene, bicyclo 2.2.1 hept-5-en-2-yl acetate, bicyclo 2.2.1 hept-5-en-2-ol, acetate, acetic acid bicyclo 2.2.1 hept-5-en-2-yl ester, 5-norbornen-2-yl acetate, mixture of endo and exo, 5-norbonen-2-yl acetate, 5-norbornen-2-ol, acetate
PubChem CID 95688
IUPAC Name 5-bicyclo[2.2.1]hept-2-enyl acetate
SMILES CC(=O)OC1CC2CC1C=C2

Specifications

Color Yellow
Boiling Point 73°C
Quantity 25 g
Formula Weight 152.19
Percent Purity ≥96.0% (GC)
Physical Form Liquid
Chemical Name or Material 5-Norbornen-2-yl Acetate (endo- and exo- mixture)