missing translation for 'onlineSavingsMsg'
Learn More

5-Nonanol 97.0+%, TCI America™

Catalog No. N033725ML
Change view
Click to view available options
Quantity:
25 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
N033725ML 25 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. N033725ML Supplier TCI America Supplier No. N033725ML

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Organic chemical building block for synthetic applications.

Chemical Identifiers

CAS 623-93-8
Molecular Formula C9H20O
Molecular Weight (g/mol) 144.258
MDL Number MFCD00021944
InChI Key FCBBRODPXVPZAH-UHFFFAOYSA-N
Synonym 5-nonanol, dibutylcarbinol, dibutyl carbinol, 5-nonyl alcohol, nonanol-5, acmc-1bc2t, di-n-butyl carbinol, 5-nonanol 25g, 5-nonanol gc, 5-nonanol, puriss gc
PubChem CID 12202
IUPAC Name nonan-5-ol
SMILES CCCCC(CCCC)O

Specifications

Melting Point 5°C
Color Yellow
Boiling Point 193°C
Quantity 25 mL
Formula Weight 144.26
Percent Purity ≥97.0% (GC)
Physical Form Liquid
Chemical Name or Material 5-Nonanol
EINECSNumber (2)-0217
TSCA No