accessibility menu, dialog, popup

UserName

H3COHCH3

CAS RN 623-93-8

IUPAC Name: nonan-5-ol
Synonyms: 5-nonanol
Molecular Weight (g/mol): 144.26
Molecular Formula: C9H20O
InChi Key: FCBBRODPXVPZAH-UHFFFAOYSA-N
SMILES: CCCCC(O)CCCC
Boiling Point 
  • (1)
  • (1)
Percent Purity 
  • (1)
  • (1)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)