missing translation for 'onlineSavingsMsg'
Learn More
Learn More
5-Nitrovanillin 98.0+%, TCI America™
$39.14 - $124.89
Chemical Identifiers
| CAS | 6635-20-7 |
|---|---|
| Molecular Formula | C8H7NO5 |
| Molecular Weight (g/mol) | 197.146 |
| MDL Number | MFCD00007118 |
| InChI Key | ZEHYRTJBFMZHCY-UHFFFAOYSA-N |
| Synonym | 5-nitrovanillin, 4-hydroxy-3-methoxy-5-nitrobenzaldehyde, vanillin, 5-nitro, benzaldehyde, 4-hydroxy-3-methoxy-5-nitro, 3-methoxy-4-hydroxy-5-nitrobenzaldhyde, 3-methoxy-4-hydroxy-5-nitrobenzaldehyde, 4-hydroxy-3-methoxy-5-nitro-benzaldehyde, 4-hydroxy-3-nitro-5-methoxybenzaldehyde, 4-hydroxy-5-methoxy-3-nitrobenzaldehyde, 5-nitrovanilline |
| PubChem CID | 81134 |
| ChEBI | CHEBI:48385 |
| SMILES | COC1=CC(=CC(=C1O)[N+](=O)[O-])C=O |
Chemical Identifiers
| 6635-20-7 | |
| 197.146 | |
| ZEHYRTJBFMZHCY-UHFFFAOYSA-N | |
| 81134 | |
| COC1=CC(=CC(=C1O)[N+](=O)[O-])C=O |
| C8H7NO5 | |
| MFCD00007118 | |
| 5-nitrovanillin, 4-hydroxy-3-methoxy-5-nitrobenzaldehyde, vanillin, 5-nitro, benzaldehyde, 4-hydroxy-3-methoxy-5-nitro, 3-methoxy-4-hydroxy-5-nitrobenzaldhyde, 3-methoxy-4-hydroxy-5-nitrobenzaldehyde, 4-hydroxy-3-methoxy-5-nitro-benzaldehyde, 4-hydroxy-3-nitro-5-methoxybenzaldehyde, 4-hydroxy-5-methoxy-3-nitrobenzaldehyde, 5-nitrovanilline | |
| CHEBI:48385 |
Specifications
| 6635-20-7 | |
| Green-Yellow | |
| MFCD00007118 | |
| 5-nitrovanillin, 4-hydroxy-3-methoxy-5-nitrobenzaldehyde, vanillin, 5-nitro, benzaldehyde, 4-hydroxy-3-methoxy-5-nitro, 3-methoxy-4-hydroxy-5-nitrobenzaldhyde, 3-methoxy-4-hydroxy-5-nitrobenzaldehyde, 4-hydroxy-3-methoxy-5-nitro-benzaldehyde, 4-hydroxy-3-nitro-5-methoxybenzaldehyde, 4-hydroxy-5-methoxy-3-nitrobenzaldehyde, 5-nitrovanilline | |
| COC1=CC(=CC(=C1O)[N+](=O)[O-])C=O | |
| 81134 | |
| 197.15 | |
| Crystalline Powder |
| 176°C | |
| C8H7NO5 | |
| 5 g | |
| ZEHYRTJBFMZHCY-UHFFFAOYSA-N | |
| 197.146 | |
| CHEBI:48385 | |
| ≥98.0% (T) | |
| 5-Nitrovanillin |
Safety and Handling
TSCA : No