missing translation for 'onlineSavingsMsg'
Learn More

5-Nitrovanillin 98.0+%, TCI America™

Catalog No. H07285G
Change view
Click to view available options
Quantity:
5 g
10 g
25 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
H07285G 5 g
H072810G 10 g
H072825G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. H07285G Supplier TCI America Supplier No. H07285G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 6635-20-7
Molecular Formula C8H7NO5
Molecular Weight (g/mol) 197.146
MDL Number MFCD00007118
InChI Key ZEHYRTJBFMZHCY-UHFFFAOYSA-N
Synonym 5-nitrovanillin, 4-hydroxy-3-methoxy-5-nitrobenzaldehyde, vanillin, 5-nitro, benzaldehyde, 4-hydroxy-3-methoxy-5-nitro, 3-methoxy-4-hydroxy-5-nitrobenzaldhyde, 3-methoxy-4-hydroxy-5-nitrobenzaldehyde, 4-hydroxy-3-methoxy-5-nitro-benzaldehyde, 4-hydroxy-3-nitro-5-methoxybenzaldehyde, 4-hydroxy-5-methoxy-3-nitrobenzaldehyde, 5-nitrovanilline
PubChem CID 81134
ChEBI CHEBI:48385
SMILES COC1=CC(=CC(=C1O)[N+](=O)[O-])C=O

Specifications

Melting Point 176°C
Color Green-Yellow
Quantity 5 g
Formula Weight 197.15
Percent Purity ≥98.0% (T)
Physical Form Crystalline Powder
Chemical Name or Material 5-Nitrovanillin