missing translation for 'onlineSavingsMsg'
Learn More

5-Nitrothiophene-2-carboxylic acid, 98%

Catalog No. p-7056075
Change view
Click to view available options
Quantity:
5 g
25 g
100 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAH6482106 5 g
AAH6482114 25 g
AAH6482122 100 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AAH6482106 Supplier Thermo Scientific Chemicals Supplier No. H6482106

CAS: 6317-37-9 | C5H3NO4S | 173.142 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 6317-37-9
Molecular Formula C5H3NO4S
Molecular Weight (g/mol) 173.142
MDL Number MFCD00159552
InChI Key UNEPVPOHGXLUIR-UHFFFAOYSA-N
Synonym 5-nitrothiophene-2-carboxylic acid, 5-nitro-2-thiophenecarboxylic acid, 5-nitro-2-thenoic acid, 5-nitrothiophene-2-carboxylicacid, 5-nitro-thiophene-2-carboxylic acid, 2-thiophenecarboxylic acid, 5-nitro, 2-carboxy-5-nitrothiophene, pubchem5568, acmc-1b47f, 5-nitrothiophene-2-carboxylic
PubChem CID 80591
SMILES C1=C(SC(=C1)[N+](=O)[O-])C(=O)O

Specifications

Melting Point 154°C to 159°C
Color Yellow
Quantity 5 g
Beilstein 139665
Formula Weight 173.14
Percent Purity 98%
Physical Form Solid
Chemical Name or Material 5-Nitrothiophene-2-carboxylic acid
Hazard Category H315-H319-H335
Hazard Statement GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
EINECSNumber 228-654-4
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only