accessibility menu, dialog, popup

UserName

CAS RN 6317-37-9

IUPAC Name: 5-nitrothiophene-2-carboxylic acid
Synonyms: 5-nitrothiophene-2-carboxylic acid
Molecular Weight (g/mol): 173.14
Molecular Formula: C5H3NO4S
InChi Key: UNEPVPOHGXLUIR-UHFFFAOYSA-N
SMILES: OC(=O)C1=CC=C(S1)[N+]([O-])=O
Percent Purity 
  • (1)
  • (3)
Physical Form 
  • (1)
  • (3)
Quantity 
  • (1)
  • (2)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (3)
  • (1)
  • (3)
  • (1)
  • (3)
  • (1)
  • (3)
  • (1)
  • (2)
  • (1)