missing translation for 'onlineSavingsMsg'
Learn More

5-Nitrosalicylic acid, 99%, Thermo Scientific™

Catalog No. AC300480050
Change view
Click to view available options
Quantity:
5g
25g
250g
Packaging:
Glass Bottle
Plastic bottle
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
AC300480050 5g Glass Bottle
AC300480250 25g Plastic bottle
AC300482500 250g Plastic bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AC300480050 Supplier Thermo Scientific Chemicals Supplier No. 300480050

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

This Thermo Scientific brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

Chemical Identifiers

CAS 96-97-9
Molecular Formula C7H5NO5
Molecular Weight (g/mol) 183.12
MDL Number MFCD00007338
InChI Key PPDRLQLKHRZIJC-UHFFFAOYSA-N
Synonym 5-nitrosalicylic acid, 2-hydroxy-5-nitrobenzoic acid, anilotic acid, benzoic acid, 2-hydroxy-5-nitro, 5-nitro-2-hydroxybenzoic acid, unii-82l9g7fyz3, 2-hydroxy-5-nitro-benzoic acid, 5-nitro-salicylic acid, salicylic acid, 5-nitro, 5-nitro salicylic acid
PubChem CID 7318
ChEBI CHEBI:61281
SMILES C1=CC(=C(C=C1[N+](=O)[O-])C(=O)O)O

Specifications

CAS Min % 98.5
CAS Max % 100.0
Infrared Spectrum Authentic
Assay Percent Range 98.5% min. (HPLC)
Packaging Glass Bottle
Linear Formula O2NC6H3(OH)COOH
Quantity 5g
Beilstein 10, 116
Merck Index 15, 6718
Solubility Information Solubility in water: 1g/1475mL in water. Other solubilities: more soluble in hot water,very soluble in ethanol,ether,acetone
Formula Weight 183.12
Percent Purity 99%
Chemical Name or Material 5-Nitrosalicylic acid, 99%
Show More Show Less
Hazard Category GHS Signal Word: Warning
Hazard Statement GHS H Statement:
Causes skin irritation.
May cause respiratory irritation.
Causes serious eye irritation.
Precautionary Statement GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
EINECSNumber 202-548-8
TSCA TSCA

RUO – Research Use Only