missing translation for 'onlineSavingsMsg'
Learn More

5-Nitrosalicylaldehyde 97.0+%, TCI America™

Catalog No. p-7133707
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
N02555G 5 g
N025525G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. N02555G Supplier TCI America Supplier No. N02555G

missing translation for 'validMainframeMsg'

Chemical Identifiers

CAS 97-51-8
Molecular Formula C7H5NO4
Molecular Weight (g/mol) 167.12
MDL Number MFCD00007337
InChI Key IHFRMUGEILMHNU-UHFFFAOYSA-N
Synonym 5-nitrosalicylaldehyde, benzaldehyde, 2-hydroxy-5-nitro, salicylaldehyde, 5-nitro, 5-nitrosaliclaldehyde, 2-hydroxy-5-nitro-benzaldehyde, 5-nitro salicylaldehdye, timtec-bb sbb003885, labotest-bb lt00920994, salicylaldehyde, 5-nitro-6ci,7ci,8ci, pubchem2052
PubChem CID 66808
IUPAC Name 2-hydroxy-5-nitrobenzaldehyde
SMILES OC1=CC=C(C=C1C=O)[N+]([O-])=O

Specifications

Melting Point 125°C
Quantity 5 g
Formula Weight 167.12
Percent Purity ≥97.0% (GC,T)
Physical Form Crystalline Powder
Chemical Name or Material 5-Nitrosalicylaldehyde
EINECSNumber (3)-1193
RTECSNumber CU6675000
TSCA Yes