missing translation for 'onlineSavingsMsg'
Learn More

5-Nitroisophthalic Acid 99.0+%, TCI America™

Catalog No. p-7133854
Change view
Click to view available options
Quantity:
25 g
500 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
N052025G 25 g
N0520500G 500 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. N052025G Supplier TCI America Supplier No. N052025G
Only null left

Chemical Identifiers

CAS 618-88-2
Molecular Formula C8H5NO6
Molecular Weight (g/mol) 211.129
MDL Number MFCD00007254
InChI Key NBDAHKQJXVLAID-UHFFFAOYSA-N
Synonym 5-nitroisophthalic acid, 5-nitrobenzene-1,3-dicarboxylic acid, 5-nitro-1,3-benzenedicarboxylic acid, 1,3-benzenedicarboxylic acid, 5-nitro, 5-nitroisophthalicacid, 5-nitro-m-phthalic acid, unii-g7rz7lc76u, isophthalic acid, 5-nitro, 1-nitrobenzene-3,5-dicarboxylic acid, g7rz7lc76u
PubChem CID 12069
SMILES C1=C(C=C(C=C1C(=O)O)[N+](=O)[O-])C(=O)O

Specifications

Melting Point 261°C
Color White-Yellow
Quantity 25 g
Formula Weight 211.13
Percent Purity ≥99.0% (HPLC)
Physical Form Crystalline Powder
Chemical Name or Material 5-Nitroisophthalic Acid