missing translation for 'onlineSavingsMsg'
Learn More
Learn More
5-Nitroisophthalic acid, 97+%
Description
5-Nitroisophthalic acid is used as an intermediate of medicine compound and as a disperser in dyes. It is used as an intermediate in organic synthesis,agrochemical,pharmaceutical and dyestuffs etc,.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
5-Nitroisophthalic acid is used as an intermediate of medicine compound and as a disperser in dyes. It is used as an intermediate in organic synthesis,agrochemical,pharmaceutical and dyestuffs etc,.
Solubility
Soluble in water.1.5 g/L at 20°C
Notes
Incompatible with strong oxidizing agents, strong bases.
5-Nitroisophthalic acid is used as an intermediate of medicine compound and as a disperser in dyes. It is used as an intermediate in organic synthesis,agrochemical,pharmaceutical and dyestuffs etc,.
Solubility
Soluble in water.1.5 g/L at 20°C
Notes
Incompatible with strong oxidizing agents, strong bases.
Specifications
Specifications
| Melting Point | 260°C to 264°C |
| Odor | Odorless |
| Quantity | 100 g |
| Beilstein | 1976587 |
| Solubility Information | Soluble in water.1.5g/L at 20°C |
| Formula Weight | 211.13 |
| Percent Purity | 98% |
| Chemical Name or Material | 5-Nitroisophthalic acid |
Safety and Handling
| Hazard Category | Warning |
| Hazard Statement | GHS H Statement H315-H319-H335 Causes skin irritation. Causes serious eye irritation. May cause respiratory irritation. |
| Precautionary Statement | GHS P Statement P280g-P305+P351+P338 Wear protective gloves. IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing. |
| EINECSNumber | 210-568-3 |
| TSCA | Yes |
| Recommended Storage | Ambient temperatures |
RUO – Research Use Only