missing translation for 'onlineSavingsMsg'
Learn More
Learn More
5-Nitroindole 98.0+%, TCI America™
$123.03 - $490.25
Chemical Identifiers
| CAS | 6146-52-7 |
|---|---|
| Molecular Formula | C8H6N2O2 |
| Molecular Weight (g/mol) | 162.148 |
| MDL Number | MFCD00005673 |
| InChI Key | OZFPSOBLQZPIAV-UHFFFAOYSA-N |
| Synonym | 5-nitroindole, 1h-indole, 5-nitro, indole, 5-nitro, unii-o2bhx6edbn, 5-nitro indole, ccris 3255, o2bhx6edbn, pubchem1711, acmc-209msr |
| PubChem CID | 22523 |
| IUPAC Name | 5-nitro-1H-indole |
| SMILES | C1=CC2=C(C=CN2)C=C1[N+](=O)[O-] |
Chemical Identifiers
| 6146-52-7 | |
| 162.148 | |
| OZFPSOBLQZPIAV-UHFFFAOYSA-N | |
| 22523 | |
| C1=CC2=C(C=CN2)C=C1[N+](=O)[O-] |
| C8H6N2O2 | |
| MFCD00005673 | |
| 5-nitroindole, 1h-indole, 5-nitro, indole, 5-nitro, unii-o2bhx6edbn, 5-nitro indole, ccris 3255, o2bhx6edbn, pubchem1711, acmc-209msr | |
| 5-nitro-1H-indole |
Specifications
| 6146-52-7 | |
| C8H6N2O2 | |
| 5 g | |
| OZFPSOBLQZPIAV-UHFFFAOYSA-N | |
| 5-nitro-1H-indole | |
| 22523 | |
| ≥98.0% (GC) | |
| 5-Nitroindole |
| 142°C | |
| MFCD00005673 | |
| 5-nitroindole, 1h-indole, 5-nitro, indole, 5-nitro, unii-o2bhx6edbn, 5-nitro indole, ccris 3255, o2bhx6edbn, pubchem1711, acmc-209msr | |
| C1=CC2=C(C=CN2)C=C1[N+](=O)[O-] | |
| 162.148 | |
| 162.15 | |
| Crystalline Powder |
Safety and Handling
EINECSNumber : (5)-6269
RTECSNumber : NM1168200
TSCA : No