missing translation for 'onlineSavingsMsg'
Learn More

5-Nitroindole 98.0+%, TCI America™

Catalog No. N04005G
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
N04005G 5 g
N040025G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. N04005G Supplier TCI America Supplier No. N04005G

Chemical Identifiers

CAS 6146-52-7
Molecular Formula C8H6N2O2
Molecular Weight (g/mol) 162.148
MDL Number MFCD00005673
InChI Key OZFPSOBLQZPIAV-UHFFFAOYSA-N
Synonym 5-nitroindole, 1h-indole, 5-nitro, indole, 5-nitro, unii-o2bhx6edbn, 5-nitro indole, ccris 3255, o2bhx6edbn, pubchem1711, acmc-209msr
PubChem CID 22523
IUPAC Name 5-nitro-1H-indole
SMILES C1=CC2=C(C=CN2)C=C1[N+](=O)[O-]

Specifications

Melting Point 142°C
Quantity 5 g
Formula Weight 162.15
Percent Purity ≥98.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material 5-Nitroindole
EINECSNumber (5)-6269
RTECSNumber NM1168200
TSCA No