missing translation for 'onlineSavingsMsg'
Learn More

5-Nitroguaiacol 98.0+%, TCI America™

Catalog No. N049825G
Change view
Click to view available options
Quantity:
25 g
500 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
N049825G 25 g
N0498500G 500 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. N049825G Supplier TCI America Supplier No. N049825G
Only null left

Chemical Identifiers

CAS 636-93-1
Molecular Formula C7H7NO4
Molecular Weight (g/mol) 169.136
MDL Number MFCD00015561
InChI Key KXKCTSZYNCDFFG-UHFFFAOYSA-N
Synonym 5-nitroguaiacol, phenol, 2-methoxy-5-nitro, 3-hydroxy-4-methoxynitrobenzene, 2-methoxy-5-nitro-phenol, unii-e5t11i2jub, e5t11i2jub, 2-hydroxy-4-nitroanisole, acmc-1b3lw, 2-methoxy-5-nitro phenol, 2-methyloxy-5-nitrophenol
PubChem CID 69471
ChEBI CHEBI:81049
IUPAC Name 2-methoxy-5-nitrophenol
SMILES COC1=C(C=C(C=C1)[N+](=O)[O-])O

Specifications

Melting Point 105°C
Boiling Point 112°C
Quantity 25 g
Formula Weight 169.14
Percent Purity ≥98.0% (T)
Physical Form Crystalline Powder
Chemical Name or Material 5-Nitroguaiacol
EINECSNumber (3)-2771&(9)-0970
TSCA No