missing translation for 'onlineSavingsMsg'
Learn More

5-Nitrobenzothiazole, 96%, Thermo Scientific Chemicals

Catalog No. AAH3359403
Change view
Click to view available options
Quantity:
1 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAH3359403 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AAH3359403 Supplier Thermo Scientific Chemicals Supplier No. H3359403
Get Up to $1,800 in Chemicals with Purchase
View all promotions

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 2942-07-6 | Molecular Formula: C12H18ClNO2 | Molecular Weight: 243.73 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 2942-07-6
MDL Number MFCD00085891

Specifications

Melting Point 157°C to 162°C
Molecular Formula C12H18ClNO2
Quantity 1 g
Synonym benzoylcholine chloride, 2-hydroxyethyl trimethylammonium chloride benzoate, choline, chloride, benzoate, 2-benzoyloxy-n,n,n-trimethylethanaminium chloride, 2-benzoyloxyethyltrimethylammonium chloride, 2-benzoyloxy ethyl trimethylazanium chloride, 2-benzyloxy-n,n,n-trimethylethanaminium chloride, 2-benzyloxy-n,n,n-trimethylethanaminimum chloride, ammonium, 2-hydroxyethyl trimethyl-, chloride, benzoate, 2-trimethylamino ethyl benzoate, chloride
InChI Key QVFHQENRNSAHEK-UHFFFAOYSA-M
SMILES C[N+](C)(C)CCOC(=O)C1=CC=CC=C1.[Cl-]
IUPAC Name 2-benzoyloxyethyl(trimethyl)azanium;chloride
Molecular Weight (g/mol) 243.73
PubChem CID 76298
Formula Weight 180.18
Percent Purity 96%
Chemical Name or Material 5-Nitrobenzothiazole
Show More Show Less
Hazard Category H315-H319-H335
Hazard Statement GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only