missing translation for 'onlineSavingsMsg'
Learn More
Learn More
5-Nitrobenzimidazole Nitrate 98.0+%, TCI America™
Supplier: TCI America N0151250G
Specifications
| 5-Nitrobenzimidazole Nitrate | |
| 222°C | |
| C7H6N4O5 | |
| 250 g | |
| C1=CC2=C(C=C1[N+](=O)[O-])NC=N2.[N+](=O)(O)[O-] | |
| 226.148 | |
| 226.15 | |
| Crystalline Powder |
| 27896-84-0 | |
| Yellow | |
| MFCD00013157 | |
| ZUZQXHSOEZUAIS-UHFFFAOYSA-N | |
| nitric acid;6-nitro-1H-benzimidazole | |
| 119743 | |
| ≥98.0% (T) |
Chemical Identifiers
| 27896-84-0 | |
| 226.148 | |
| ZUZQXHSOEZUAIS-UHFFFAOYSA-N | |
| nitric acid;6-nitro-1H-benzimidazole |
| C7H6N4O5 | |
| MFCD00013157 | |
| 119743 | |
| C1=CC2=C(C=C1[N+](=O)[O-])NC=N2.[N+](=O)(O)[O-] |
Safety and Handling
RTECSNumber : DD9801000
TSCA : Yes