missing translation for 'onlineSavingsMsg'
Learn More

5-Nitroacenaphthene (contains 3-Nitroacenaphthene) 80.0+%, TCI America™

Catalog No. N01055G
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
N01055G 5 g
N010525G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. N01055G Supplier TCI America Supplier No. N01055G
Only null left

Chemical Identifiers

CAS 602-87-9
Molecular Formula C12H9NO2
Molecular Weight (g/mol) 199.21
MDL Number MFCD00014277
InChI Key CUARLQDWYSRQDF-UHFFFAOYSA-N
Synonym 5-nitroacenaphthene, 5-nitroacenapthene, acenaphthene, 5-nitro, acenaphthylene, 1,2-dihydro-5-nitro, 5-nan, 1,2-dihydro-5-nitro-acenaphthylene, 5-nitronaphthalene ethylene, 5-nitronaphthalene, 1,2-dihydro-5-nitroacenaphthylene, ccris 438
PubChem CID 11769
ChEBI CHEBI:82350
IUPAC Name 5-nitro-1,2-dihydroacenaphthylene
SMILES [O-][N+](=O)C1=C2C=CC=C3CCC(C=C1)=C23

Specifications

Melting Point 102°C
Color Yellow
Quantity 5 g
UN Number 2811
Formula Weight 199.21
Percent Purity ≥80.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material 5-Nitroacenaphthene (contains 3-Nitroacenaphthene)
EINECSNumber (4)-0352
RTECSNumber AB1060000
TSCA Yes