missing translation for 'onlineSavingsMsg'
Learn More

5-Nitro-2-thiophenecarboxylic Acid 97.0+%, TCI America™

Catalog No. p-7134037
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
N09405G 5 g
N094025G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. N09405G Supplier TCI America Supplier No. N09405G
Only null left

Chemical Identifiers

CAS 6317-37-9
Molecular Formula C5H3NO4S
Molecular Weight (g/mol) 173.142
MDL Number MFCD00159552
InChI Key UNEPVPOHGXLUIR-UHFFFAOYSA-N
Synonym 5-nitrothiophene-2-carboxylic acid, 5-nitro-2-thiophenecarboxylic acid, 5-nitro-2-thenoic acid, 5-nitrothiophene-2-carboxylicacid, 5-nitro-thiophene-2-carboxylic acid, 2-thiophenecarboxylic acid, 5-nitro, 2-carboxy-5-nitrothiophene, pubchem5568, acmc-1b47f, 5-nitrothiophene-2-carboxylic
PubChem CID 80591
SMILES C1=C(SC(=C1)[N+](=O)[O-])C(=O)O

Specifications

Melting Point 160°C
Quantity 5 g
Formula Weight 173.14
Percent Purity ≥97.0% (GC,T)
Physical Form Crystalline Powder
Chemical Name or Material 5-Nitro-2-thiophenecarboxylic Acid