missing translation for 'onlineSavingsMsg'
Learn More

5-Nitro-2-benzimidazolinone, 99%

Catalog No. AAL1639106
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAL1639106 5 g
AAL1639114 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAL1639106 Supplier Thermo Scientific Chemicals Supplier No. L1639106
Get Up to $1,800 in Chemicals with Purchase
View all promotions

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 93-84-5 | C7H5N3O3 | 179.14 g/mol

5-Nitro-2-benzimidazolinone is used as intermediates.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
5-Nitro-2-benzimidazolinone is used as intermediates.

Solubility
Insoluble in water.

Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Store away from strong oxidizing agents.

Chemical Identifiers

CAS 93-84-5
Molecular Formula C7H5N3O3
Molecular Weight (g/mol) 179.14
MDL Number MFCD00220274
InChI Key DLJZIPVEVJOKHB-UHFFFAOYSA-N
Synonym 5-nitro-2-benzimidazolinone, 5-nitro-1h-benzo d imidazol-2 3h-one, 5-nitrobenzimidazol-2-one, 5-nitro-1,3-dihydro-2h-benzimidazol-2-one, 2-hydroxy-5-nitrobenzimidazole, 5-nitro-2 3h-benzimidazolone, 2h-benzimidazol-2-one, 1,3-dihydro-5-nitro, 2-benzimidazolinone, 5-nitro, 5-nitro-1h-benzimidazol-2-ol, 1,3-dihydro-5-nitro-2h-benzimidazol-2-one
PubChem CID 3725611
SMILES [O-][N+](=O)C1=CC=C2NC(=O)NC2=C1

Specifications

Melting Point >300°C
Quantity 5 g
UN Number UN2811
Beilstein 171386
Solubility Information Insoluble in water.
IUPAC Name 5-nitro-1,3-dihydrobenzimidazol-2-one
Formula Weight 179.14
Percent Purity 99%
Chemical Name or Material 5-Nitro-2-benzimidazolinone
Hazard Category H302+H312+H332-H319
Hazard Statement GHS H Statement
H331-H302
Toxic if inhaled.
Harmful if swallowed.
Precautionary Statement P261-P264b-P270-P271-P280-P301+P312-P302+P352-P304+P340-P305+P351+P338-P312-P330-P363-P501c
DOTInformation Transport Hazard Class: 6.1; Packing Group: III; Proper Shipping Name: TOXIC SOLIDS, ORGANIC, N.O.S.
EINECSNumber 202-282-2
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only