accessibility menu, dialog, popup

UserName

CAS RN 93-84-5

IUPAC Name: 5-nitro-2,3-dihydro-1H-1,3-benzodiazol-2-one
Synonyms: 5-nitro-1,3-dihydro-1,3-benzodiazol-2-one
Molecular Weight (g/mol): 179.14
Molecular Formula: C7H5N3O3
InChi Key: DLJZIPVEVJOKHB-UHFFFAOYSA-N
SMILES: [O-][N+](=O)C1=CC=C2NC(=O)NC2=C1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results