missing translation for 'onlineSavingsMsg'
Learn More

5-Nitro-1H-indazole, 98+%

Catalog No. AAA1347814
Change view
Click to view available options
Quantity:
25 g
100 g
500 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAA1347814 25 g
AAA1347822 100 g
AAA1347836 500 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AAA1347814 Supplier Thermo Scientific Chemicals Supplier No. A1347814
Get Up to $1,800 in Chemicals with Purchase
View all promotions

CAS: 5401-94-5 | C7H5N3O2 | 163.136 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 5401-94-5
Molecular Formula C7H5N3O2
Molecular Weight (g/mol) 163.136
MDL Number MFCD00005693
InChI Key WSGURAYTCUVDQL-UHFFFAOYSA-N
Synonym 5-nitroindazole, 1h-indazole, 5-nitro, unii-235y7p37zd, ccris 4134, 2h-indazole, 5-nitro, 5ni, 5-nitroindazol, 5-nitro indazole, 5-nitro-indazole, 5-nitro-1h-indazol
PubChem CID 21501
IUPAC Name 5-nitro-1H-indazole
SMILES C1=CC2=C(C=C1[N+](=O)[O-])C=NN2

Specifications

Melting Point 208°C to 211°C
Odor Odorless
Quantity 25 g
Beilstein 7936
Formula Weight 163.14
Percent Purity ≥98%
Chemical Name or Material 5-Nitro-1H-indazole
Hazard Category Warning
Hazard Statement GHS H Statement
H341-H315-H319-H335
Suspected of causing genetic defects.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement GHS P Statement
P261-P280-P281-P305+P351+P338-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
Use personal protective equipment as required.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
EINECSNumber 226-451-5
RTECSNumber NK7962000
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only