missing translation for 'onlineSavingsMsg'
Learn More
Learn More
5-Methyluridine 98.0+%, TCI America™
$39.14 - $160.30
Chemical Identifiers
| CAS | 1463-10-1 |
|---|---|
| Molecular Formula | C10H14N2O6 |
| Molecular Weight (g/mol) | 258.23 |
| MDL Number | MFCD00006535 |
| InChI Key | DWRXFEITVBNRMK-JXOAFFINSA-N |
| Synonym | 5-methyluridine, ribothymidine, thymine riboside, ribosylthymine, uridine, 5-methyl, ribosylthymidine, 5-methyl-uridine, beta-d-ribofuranoside, thymine ribofuranoside, 1-beta-d-ribofuranosyl thymine |
| PubChem CID | 445408 |
| ChEBI | CHEBI:45996 |
| IUPAC Name | 1-[(2R,3R,4S,5R)-3,4-dihydroxy-5-(hydroxymethyl)oxolan-2-yl]-5-methylpyrimidine-2,4-dione |
| SMILES | CC1=CN(C(=O)NC1=O)C2C(C(C(O2)CO)O)O |
Chemical Identifiers
| 1463-10-1 | |
| 258.23 | |
| DWRXFEITVBNRMK-JXOAFFINSA-N | |
| 445408 | |
| 1-[(2R,3R,4S,5R)-3,4-dihydroxy-5-(hydroxymethyl)oxolan-2-yl]-5-methylpyrimidine-2,4-dione |
| C10H14N2O6 | |
| MFCD00006535 | |
| 5-methyluridine, ribothymidine, thymine riboside, ribosylthymine, uridine, 5-methyl, ribosylthymidine, 5-methyl-uridine, beta-d-ribofuranoside, thymine ribofuranoside, 1-beta-d-ribofuranosyl thymine | |
| CHEBI:45996 | |
| CC1=CN(C(=O)NC1=O)C2C(C(C(O2)CO)O)O |
Specifications
| 1463-10-1 | |
| White | |
| MFCD00006535 | |
| 5-methyluridine, ribothymidine, thymine riboside, ribosylthymine, uridine, 5-methyl, ribosylthymidine, 5-methyl-uridine, beta-d-ribofuranoside, thymine ribofuranoside, 1-beta-d-ribofuranosyl thymine | |
| CC1=CN(C(=O)NC1=O)C2C(C(C(O2)CO)O)O | |
| 258.23 | |
| CHEBI:45996 | |
| ≥98.0% (HPLC,T) | |
| 5-Methyluridine |
| 183°C | |
| C10H14N2O6 | |
| 1 g | |
| DWRXFEITVBNRMK-JXOAFFINSA-N | |
| 1-[(2R,3R,4S,5R)-3,4-dihydroxy-5-(hydroxymethyl)oxolan-2-yl]-5-methylpyrimidine-2,4-dione | |
| 445408 | |
| 258.23 | |
| Crystalline Powder |
Safety and Handling
EINECSNumber : (5)-6843
TSCA : No