missing translation for 'onlineSavingsMsg'
Learn More

5-Methyluridine, 99%

CAS: 1463-10-1 | C10H14N2O6 | 258.23 g/mol

$149.70 - $430.74

Chemical Identifiers

CAS 1463-10-1
Molecular Formula C10H14N2O6
Molecular Weight (g/mol) 258.23
MDL Number MFCD00006535
InChI Key DWRXFEITVBNRMK-JXOAFFINSA-N
Synonym 5-methyluridine, ribothymidine, thymine riboside, ribosylthymine, uridine, 5-methyl, ribosylthymidine, 5-methyl-uridine, beta-d-ribofuranoside, thymine ribofuranoside, 1-beta-d-ribofuranosyl thymine
PubChem CID 445408
ChEBI CHEBI:45996
IUPAC Name 1-[(2R,3R,4S,5R)-3,4-dihydroxy-5-(hydroxymethyl)oxolan-2-yl]-5-methylpyrimidine-2,4-dione
SMILES CC1=CN(C(=O)NC1=O)C2C(C(C(O2)CO)O)O
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
AAH6425914
View Documents
Thermo Scientific Chemicals
H6425914
25 g
Each for $149.70
Only null left
 
AAH6425922
View Documents
Thermo Scientific Chemicals
H6425922
100 g
Each for $430.74
Only null left
 
Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Chemical Identifiers

1463-10-1
258.23
DWRXFEITVBNRMK-JXOAFFINSA-N
445408
1-[(2R,3R,4S,5R)-3,4-dihydroxy-5-(hydroxymethyl)oxolan-2-yl]-5-methylpyrimidine-2,4-dione
C10H14N2O6
MFCD00006535
5-methyluridine, ribothymidine, thymine riboside, ribosylthymine, uridine, 5-methyl, ribosylthymidine, 5-methyl-uridine, beta-d-ribofuranoside, thymine ribofuranoside, 1-beta-d-ribofuranosyl thymine
CHEBI:45996
CC1=CN(C(=O)NC1=O)C2C(C(C(O2)CO)O)O

Specifications

1463-10-1
White
MFCD00006535
90164
DWRXFEITVBNRMK-JXOAFFINSA-N
1-[(2R,3R,4S,5R)-3,4-dihydroxy-5-(hydroxymethyl)oxolan-2-yl]-5-methylpyrimidine-2,4-dione
445408
258.23
Solid
183°C to 184°C
C10H14N2O6
25 g
5-methyluridine, ribothymidine, thymine riboside, ribosylthymine, uridine, 5-methyl, ribosylthymidine, 5-methyl-uridine, beta-d-ribofuranoside, thymine ribofuranoside, 1-beta-d-ribofuranosyl thymine
CC1=CN(C(=O)NC1=O)C2C(C(C(O2)CO)O)O
258.23
CHEBI:45996
99%
5-Methyluridine
Videos
Safety and Handling

Safety and Handling

EINECSNumber : 215-973-9

TSCA : No

storageNote1 : 2° to 8°C

Recommended Storage : Keep cold

SDS
missing translation for 'documents'

missing translation for 'documents'

RUO – Research Use Only