missing translation for 'onlineSavingsMsg'
Learn More

5-Methylthiophene-2-sulfonyl chloride, 97%, Thermo Scientific™

Supplier:  Thermo Scientific Chemicals CC65003DE

Quantity 5g

 View more versions of this product

Catalog No. CC65003DE

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Description

Description

Specifications

Chemical Identifiers

55854-45-0
196.66
QGGRRHYGHGJEKP-UHFFFAOYSA-N
12400112
CC1=CC=C(S1)S(Cl)(=O)=O
C5H5ClO2S2
MFCD05864499
5-methyl-2-thiophenesulfonyl chloride, 2-thiophenesulfonyl chloride, 5-methyl, 5-methylthiophene-2-sulfonylchloride, 5-methylthiophene-2-sulphonyl chloride, 5-methyl-2-thienylsulfonyl chloride, chloro 5-methyl 2-thienyl sulfone, acmc-209lpu, 5-methylthiophenesulphonyl chloride, 5-methyl-thiophenesulfonyl chloride, 2-chlorosulphonyl-5-methylthiophene
5-methylthiophene-2-sulfonyl chloride

Specifications

5-Methylthiophene-2-sulfonyl chloride
96.5
95%
C5H5ClO2S2
MFCD05864499
QGGRRHYGHGJEKP-UHFFFAOYSA-N
5-methylthiophene-2-sulfonyl chloride
12400112
97%
55854-45-0
100.0
Amber Glass Bottle
5g
5-methyl-2-thiophenesulfonyl chloride, 2-thiophenesulfonyl chloride, 5-methyl, 5-methylthiophene-2-sulfonylchloride, 5-methylthiophene-2-sulphonyl chloride, 5-methyl-2-thienylsulfonyl chloride, chloro 5-methyl 2-thienyl sulfone, acmc-209lpu, 5-methylthiophenesulphonyl chloride, 5-methyl-thiophenesulfonyl chloride, 2-chlorosulphonyl-5-methylthiophene
CC1=CC=C(S1)S(Cl)(=O)=O
196.66
196.68
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

GHS H Statement:
Causes serious eye damage.
Causes severe skin burns and eye damage.
Contact with water liberates toxic gas.
Reacts violently with water.

GHS P Statement:
IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
Wear eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Immediately call a POISON CENTER or doctor/physician.
Store in a dry place. Store in a closed container.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS Signal Word: Danger

SDS
Documents

Documents