missing translation for 'onlineSavingsMsg'
Learn More

5-Methylthiophene-2-sulfonyl chloride, 97%, Thermo Scientific™

Catalog No. p-3541135
Change view
Click to view available options
Quantity:
1g
5g
10g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
CC65003DA 1g
CC65003DE 5g
CC65003EA 10g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. CC65003DA Supplier Thermo Scientific Chemicals Supplier No. CC65003DA

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 55854-45-0
Molecular Formula C5H5ClO2S2
Molecular Weight (g/mol) 196.66
MDL Number MFCD05864499
InChI Key QGGRRHYGHGJEKP-UHFFFAOYSA-N
Synonym 5-methyl-2-thiophenesulfonyl chloride, 2-thiophenesulfonyl chloride, 5-methyl, 5-methylthiophene-2-sulfonylchloride, 5-methylthiophene-2-sulphonyl chloride, 5-methyl-2-thienylsulfonyl chloride, chloro 5-methyl 2-thienyl sulfone, acmc-209lpu, 5-methylthiophenesulphonyl chloride, 5-methyl-thiophenesulfonyl chloride, 2-chlorosulphonyl-5-methylthiophene
PubChem CID 12400112
IUPAC Name 5-methylthiophene-2-sulfonyl chloride
SMILES CC1=CC=C(S1)S(Cl)(=O)=O

Specifications

CAS Min % 96.5
CAS Max % 100.0
Assay Percent Range 95%
Packaging Amber Glass Bottle
Quantity 1g
Formula Weight 196.68
Percent Purity 97%
Chemical Name or Material 5-Methylthiophene-2-sulfonyl chloride
Hazard Category GHS Signal Word: Danger
Hazard Statement GHS H Statement:
Causes serious eye damage.
Causes severe skin burns and eye damage.
Contact with water liberates toxic gas.
Reacts violently with water.
Precautionary Statement GHS P Statement:
IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
Wear eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Immediately call a POISON CENTER or doctor/physician.
Store in a dry place. Store in a closed container.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.