missing translation for 'onlineSavingsMsg'
Learn More

5-(Methylsulfonyl)pyridine-3-boronic acid, 98%

Catalog No. AAH53133MD
Change view
Click to view available options
Quantity:
250 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAH53133MD 250 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AAH53133MD Supplier Thermo Scientific Chemicals Supplier No. H53133MD
Get Up to $1,800 in Chemicals with Purchase
View all promotions

CAS: 913836-01-8 | C6H8BNO4S | 201.00 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 913836-01-8
Molecular Formula C6H8BNO4S
Molecular Weight (g/mol) 201.00
MDL Number MFCD08689536
InChI Key YGXNPQDXWDCWET-UHFFFAOYSA-N
Synonym 5-methylsulphonyl pyridine-3-boronic acid, 5-methylsulfonyl pyridine-3-boronic acid, 5-methylsulfonyl pyridin-3-yl boronic acid, 5-methylsulfonyl-3-pyridineboronic acid, boronicacid, b-5-methylsulfonyl-3-pyridinyl, 5-methanesulfonylpyridin-3-ylboronic acid, b-5-methylsulfonyl-3-pyridinyl boronic acid, acmc-209ram
PubChem CID 44119381
IUPAC Name (5-methanesulfonylpyridin-3-yl)boronic acid
SMILES CS(=O)(=O)C1=CN=CC(=C1)B(O)O

Specifications

Melting Point 123°C to 125°C
Quantity 250 mg
Formula Weight 201
Percent Purity 98%
Chemical Name or Material 5-(Methylsulfonyl)pyridine-3-boronic acid
Hazard Category H315-H319-H335
Hazard Statement GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
TSCA No
Recommended Storage Keep cold

RUO – Research Use Only