missing translation for 'onlineSavingsMsg'
Learn More

5-Methylquinoxaline 98.0+%, TCI America™

Catalog No. M13711G
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
M13711G 1 g
M13715G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. M13711G Supplier TCI America Supplier No. M13711G

Chemical Identifiers

CAS 13708-12-8
Molecular Formula C9H8N2
Molecular Weight (g/mol) 144.177
MDL Number MFCD00012335
InChI Key CQLOYHZZZCWHSG-UHFFFAOYSA-N
Synonym quinoxaline, 5-methyl, unii-l0ad7t152g, 5-methyl quinoxaline, fema no. 3203, 8-methylquinoxaline, 5-methyl-quinoxaline, acmc-209caq, 5-methylquinoxaline, dsstox_cid_27090
PubChem CID 61670
IUPAC Name 5-methylquinoxaline
SMILES CC1=C2C(=CC=C1)N=CC=N2

Specifications

Boiling Point 120°C
Quantity 1 g
Formula Weight 144.18
Percent Purity ≥98.0% (GC)
Physical Form Liquid
Chemical Name or Material 5-Methylquinoxaline