accessibility menu, dialog, popup

UserName

CH3NN

CAS RN 13708-12-8

IUPAC Name: 5-methylquinoxaline
Synonyms: 5-methylquinoxaline
Molecular Weight (g/mol): 144.18
Molecular Formula: C9H8N2
InChi Key: CQLOYHZZZCWHSG-UHFFFAOYSA-N
SMILES: CC1=C2N=CC=NC2=CC=C1
Boiling Point 
  • (2)
  • (2)
Percent Purity 
  • (2)
  • (2)
Quantity 
  • (2)
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (4)
  • (2)
  • (2)