missing translation for 'onlineSavingsMsg'
Learn More

5-Methylquinoxaline, 98%

Catalog No. p-7024511
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAA1930003 1 g
AAA1930006 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAA1930003 Supplier Thermo Scientific Chemicals Supplier No. A1930003

CAS: 13708-12-8 | C9H8N2 | 144.177 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 13708-12-8
Molecular Formula C9H8N2
Molecular Weight (g/mol) 144.177
MDL Number MFCD00012335
InChI Key CQLOYHZZZCWHSG-UHFFFAOYSA-N
Synonym quinoxaline, 5-methyl, unii-l0ad7t152g, 5-methyl quinoxaline, fema no. 3203, 8-methylquinoxaline, 5-methyl-quinoxaline, acmc-209caq, 5-methylquinoxaline, dsstox_cid_27090
PubChem CID 61670
IUPAC Name 5-methylquinoxaline
SMILES CC1=C2C(=CC=C1)N=CC=N2

Specifications

Melting Point 20°C to 21°C
Density 1.12
Boiling Point 119°C to 121°C (15 mmHg)
Flash Point >110°C (230°F)
Refractive Index 1.62
Quantity 1 g
Formula Weight 144.18
Percent Purity 98%
Chemical Name or Material 5-Methylquinoxaline
Hazard Category H315-H319-H335
Hazard Statement GHS H Statement
H315-H319
Causes skin irritation.
Causes serious eye irritation.
Precautionary Statement P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
EINECSNumber 237-246-5
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only