missing translation for 'onlineSavingsMsg'
Learn More

5-Methylpyridine-2-carbonitrile, 97+%

Catalog No. p-7055853
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAH6453906 5 g
AAH6453914 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAH6453906 Supplier Thermo Scientific Chemicals Supplier No. H6453906
Only null left

CAS: 1620-77-5 | C7H6N2 | 118.14 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 1620-77-5
Molecular Formula C7H6N2
Molecular Weight (g/mol) 118.14
MDL Number MFCD06200830
InChI Key LIEQVZZZYLHNRH-UHFFFAOYSA-N
Synonym 2-cyano-5-methylpyridine, 5-methylpicolinonitrile, 5-methyl-pyridine-2-carbonitrile, 5-methyl-2-pyridinecarbonitrile, 6-cyano-3-picoline, 2-pyridinecarbonitrile, 5-methyl, 2-cyano-5-methylpyridine 2-cyano-5-picoline, 5-methypicolinonitrile, pubchem12978, 5-methyl picolinonitrile
PubChem CID 74183
IUPAC Name 5-methylpyridine-2-carbonitrile
SMILES CC1=CC=C(N=C1)C#N

Specifications

Melting Point 72°C to 75°C
Quantity 5 g
UN Number UN3439
Beilstein 111125
Formula Weight 118.14
Percent Purity ≥97%
Physical Form Solid
Chemical Name or Material 5-Methylpyridine-2-carbonitrile
Hazard Category H302+H312+H332-H315-H319-H335
Hazard Statement GHS H Statement
H331-H302-H312-H315-H319-H335
Toxic if inhaled.
Harmful if swallowed.
Harmful in contact with skin.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P270-P271-P280-P301+P312-P302+P352-P304+P340-P305+P351+P338-P312-P330-P332+P313-P362-P501c
DOTInformation Transport Hazard Class: 6.1; Packaging Group: III; Proper Shipping Name: NITRILES, SOLID, TOXIC, N.O.S.
EINECSNumber 216-589-4
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only