accessibility menu, dialog, popup

UserName

CH3NN

CAS RN 1620-77-5

IUPAC Name: 5-methylpyridine-2-carbonitrile
Synonyms: 5-methylpyridine-2-carbonitrile
Molecular Weight (g/mol): 118.14
Molecular Formula: C7H6N2
InChi Key: LIEQVZZZYLHNRH-UHFFFAOYSA-N
SMILES: CC1=CC=C(N=C1)C#N
Percent Purity 
  • (2)
  • (2)
Physical Form 
  • (2)
  • (2)
Quantity 
  • (1)
  • (1)
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (1)
  • (1)
  • (2)
  • (2)