missing translation for 'onlineSavingsMsg'
Learn More

5-Methylpyridine-2-boronic acid, 95%

Catalog No. AAH52517MD
Change view
Click to view available options
Quantity:
250 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAH52517MD 250 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AAH52517MD Supplier Thermo Scientific Chemicals Supplier No. H52517MD
Get Up to $1,800 in Chemicals with Purchase
View all promotions

CAS: 372963-49-0 | C6H8BNO2 | 136.945 g/mol

5-Methylpyridine-2-boronic acid is used as an organic chemical synthesis intermediate.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
5-Methylpyridine-2-boronic acid is used as an organic chemical synthesis intermediate.

Solubility
Slightly soluble in water.

Notes
Stable under recommended storage conditions. Incompatible with oxid

Chemical Identifiers

CAS 372963-49-0
Molecular Formula C6H8BNO2
Molecular Weight (g/mol) 136.945
MDL Number MFCD00674179
InChI Key VDRIXWWGBDNWCM-UHFFFAOYSA-N
Synonym 5-methylpyridine-2-boronic acid, 5-methylpyridin-2-yl boronic acid, 5-methyl-2-pyridineboronic acid, 5-picoline-2-boronic acid, boronic acid, 5-methyl-2-pyridinyl, 5-methyl-2-pyridinyl-boronic acid, pubchem6938, acmc-209is4, 5-methyl-2-pyridyl boronic acid, 5-methyl-2-pyridinyl boronic acid
PubChem CID 2762743
IUPAC Name (5-methylpyridin-2-yl)boronic acid
SMILES B(C1=NC=C(C=C1)C)(O)O

Specifications

Melting Point 86°C to 90°C
Quantity 250 mg
Solubility Information Slightly soluble in water.
Formula Weight 136.95
Percent Purity 95%
Chemical Name or Material 5-Methylpyridine-2-boronic acid
Hazard Category H315-H319-H335
Hazard Statement GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
TSCA No
Recommended Storage Store at -20°C

RUO – Research Use Only