CAS RN 372963-49-0
IUPAC Name:
(5-methylpyridin-2-yl)boronic acid
Synonyms:
5-methylpyridin-2-ylboronic acid
Molecular Weight (g/mol):
136.95
Molecular Formula:
C6H8BNO2
InChi Key:
VDRIXWWGBDNWCM-UHFFFAOYSA-N
SMILES:
CC1=CC=C(N=C1)B(O)O
1
–
1
of
1
results
1
Promotions Available
5-Methylpyridine-2-boronic acid, 95%
5-Methylpyridine-2-boronic acid, 95%
CAS: 372963-49-0 Molecular Formula: C6H8BNO2 Molecular Weight (g/mol): 136.945 MDL Number: MFCD00674179 InChI Key: VDRIXWWGBDNWCM-UHFFFAOYSA-N Synonym: 5-methylpyridine-2-boronic acid,5-methylpyridin-2-yl boronic acid,5-methyl-2-pyridineboronic acid,5-picoline-2-boronic acid,boronic acid, 5-methyl-2-pyridinyl,5-methyl-2-pyridinyl-boronic acid,pubchem6938,acmc-209is4,5-methyl-2-pyridyl boronic acid,5-methyl-2-pyridinyl boronic acid PubChem CID: 2762743 IUPAC Name: (5-methylpyridin-2-yl)boronic acid SMILES: B(C1=NC=C(C=C1)C)(O)O