missing translation for 'onlineSavingsMsg'
Learn More

5-Methylnicotinamide, 97%

Catalog No. AAL1897006
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAL1897006 5 g
AAL1897003 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAL1897006 Supplier Thermo Scientific Chemicals Supplier No. L1897006
Get Up to $1,800 in Chemicals with Purchase
View all promotions

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 70-57-5 | C7H8N2O | 136.154 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 70-57-5
Molecular Formula C7H8N2O
Molecular Weight (g/mol) 136.154
MDL Number MFCD05664041
InChI Key BCCUXBGEPLKSEX-UHFFFAOYSA-N
PubChem CID 65560
IUPAC Name 5-methylpyridine-3-carboxamide
SMILES CC1=CN=CC(=C1)C(=O)N

Specifications

Melting Point 166°C to 168°C
Quantity 5 g
Beilstein 4962976
Synonym 5-methylnicotinamide, nicotinamide, 5-methyl, 5-methyl-3-pyridinecarboxamide, 3-pyridinecarboxamide, 5-methyl-9ci, 5-methyl nicotinamide, 3-pyridinecarboxamide,5-methyl, 3-pyridinecarboxamide, 5-methyl
Formula Weight 136.15
Percent Purity 97%
Chemical Name or Material 5-Methylnicotinamide
Hazard Category H315-H319-H335
Hazard Statement GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
EINECSNumber 000-000-0
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only