accessibility menu, dialog, popup

UserName

CH3NNH2O

CAS RN 70-57-5

IUPAC Name: 5-methylpyridine-3-carboxamide
Synonyms: 5-methylpyridine-3-carboxamide
Molecular Weight (g/mol): 136.15
Molecular Formula: C7H8N2O
InChi Key: BCCUXBGEPLKSEX-UHFFFAOYSA-N
SMILES: CC1=CN=CC(=C1)C(N)=O

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results