missing translation for 'onlineSavingsMsg'
Learn More

5-Methylisoxazole-3-carboxylic Acid 98.0+%, TCI America™

Catalog No. p-7133207
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
M24055G 5 g
M240525G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. M24055G Supplier TCI America Supplier No. M24055G
Only null left

Chemical Identifiers

CAS 3405-77-4
Molecular Formula C5H5NO3
Molecular Weight (g/mol) 127.099
MDL Number MFCD01318162
InChI Key BNMPIJWVMVNSRD-UHFFFAOYSA-N
Synonym 5-methylisoxazole-3-carboxylic acid, 5-methylisoxazole-3-carboxylicacid, 3-isoxazolecarboxylic acid, 5-methyl, 5-methyl-3-isoxazolecarboxylic acid, 5-methyl-isoxazole-3-carboxylic acid, 3-carboxy-5-methylisoxazole, 3-isoxazolecarboxylicacid, 5-methyl, isoxazole acid, pubchem15552, ksc224c5p
PubChem CID 76947
IUPAC Name 5-methyl-1,2-oxazole-3-carboxylic acid
SMILES CC1=CC(=NO1)C(=O)O

Specifications

Melting Point 174°C
Color White-Yellow
Quantity 5 g
Formula Weight 127.10
Percent Purity ≥98.0% (HPLC,T)
Physical Form Crystalline Powder
Chemical Name or Material 5-Methylisoxazole-3-carboxylic Acid