accessibility menu, dialog, popup

UserName

CAS RN 3405-77-4

IUPAC Name: 5-methyl-1,2-oxazole-3-carboxylic acid
Synonyms: 5-methyl-1,2-oxazole-3-carboxylic acid
Molecular Weight (g/mol): 127.1
Molecular Formula: C5H5NO3
InChi Key: BNMPIJWVMVNSRD-UHFFFAOYSA-N
SMILES: CC1=CC(=NO1)C(O)=O
Percent Purity 
  • (2)
  • (2)
  • (1)
Quantity 
  • (1)
  • (2)
  • (2)
Physical Form 
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (2)
  • (3)
  • (2)
  • (2)
  • (1)
  • (1)
  • (2)
  • (2)
  • (2)