missing translation for 'onlineSavingsMsg'
Learn More

5-Methylisoxazole-3-carboxylic acid, 97%, Thermo Scientific™

$22.56 - $22.56

Chemical Identifiers

CAS 3405-77-4
Molecular Formula C5H5NO3
Molecular Weight (g/mol) 127.099
MDL Number MFCD01318162
InChI Key BNMPIJWVMVNSRD-UHFFFAOYSA-N
Synonym 5-methylisoxazole-3-carboxylic acid, 5-methylisoxazole-3-carboxylicacid, 3-isoxazolecarboxylic acid, 5-methyl, 5-methyl-3-isoxazolecarboxylic acid, 5-methyl-isoxazole-3-carboxylic acid, 3-carboxy-5-methylisoxazole, 3-isoxazolecarboxylicacid, 5-methyl, isoxazole acid, pubchem15552, ksc224c5p
PubChem CID 76947
IUPAC Name 5-methyl-1,2-oxazole-3-carboxylic acid
SMILES CC1=CC(=NO1)C(=O)O
View More Specs

Products 1
Catalog Number Mfr. No. Quantity Packaging Price Quantity  
Catalog Number Mfr. No. Quantity Packaging Price Quantity  
CC10401DA
SDS View Documents
Thermo Scientific Chemicals
CC10401DA
1 g Amber Glass Bottle
Each for $22.56
Only null left
 
Description

Description

Specifications

Chemical Identifiers

3405-77-4
127.099
BNMPIJWVMVNSRD-UHFFFAOYSA-N
76947
CC1=CC(=NO1)C(=O)O
C5H5NO3
MFCD01318162
5-methylisoxazole-3-carboxylic acid, 5-methylisoxazole-3-carboxylicacid, 3-isoxazolecarboxylic acid, 5-methyl, 5-methyl-3-isoxazolecarboxylic acid, 5-methyl-isoxazole-3-carboxylic acid, 3-carboxy-5-methylisoxazole, 3-isoxazolecarboxylicacid, 5-methyl, isoxazole acid, pubchem15552, ksc224c5p
5-methyl-1,2-oxazole-3-carboxylic acid

Specifications

3405-77-4
100.0
Amber Glass Bottle
MFCD01318162
5-methylisoxazole-3-carboxylic acid, 5-methylisoxazole-3-carboxylicacid, 3-isoxazolecarboxylic acid, 5-methyl, 5-methyl-3-isoxazolecarboxylic acid, 5-methyl-isoxazole-3-carboxylic acid, 3-carboxy-5-methylisoxazole, 3-isoxazolecarboxylicacid, 5-methyl, isoxazole acid, pubchem15552, ksc224c5p
CC1=CC(=NO1)C(=O)O
127.099
127.1
5-Methylisoxazole-3-carboxylic acid
96.5
97%
C5H5NO3
1 g
BNMPIJWVMVNSRD-UHFFFAOYSA-N
5-methyl-1,2-oxazole-3-carboxylic acid
76947
97%
Videos
Safety and Handling

Safety and Handling

GHS H Statement:
May cause respiratory irritation.
Causes skin irritation.
Causes serious eye irritation.

GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

SDS
missing translation for 'documents'

missing translation for 'documents'