Learn More
5-Methylimidazole-4-carboxaldehyde, 99%, Thermo Scientific Chemicals
Description
4-Methyl-5-imidazolecarboxaldehyde is suitable for use in the synthesis of 1,3-bis[(4-methyl-5-imidazol-1-yl)ethylideneamino]-propan-2-ol (BIPO), 1,3-bis[(4-methyl-5-imidazol-1-yl)ethylideneamino]propane (BIP), {acetatoaqua[[(4-methylimidazol-5-yl)methylene]histamine]-copper(II)} perchlorate, Schiff base condensates with tris(2-aminoethyl)amine (tren) or tris(3-aminopropyl)amine (trpn). It is also used in the synthesis of imidazolate-based metal organic framework (MOF) membrane, 3,11-bis(4-imidazolylmethyl)-7-methyl-3,7,11,17-tetraazabicyclo[11.3.1]hepta-deca-1(17),13,15-triene, a 4-imidazolyl derivative of a 14-membered tetraazamacrocycle and Cu(II) eight and six coordinate compounds.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
4-Methyl-5-imidazolecarboxaldehyde is suitable for use in the synthesis of 1,3-bis[(4-methyl-5-imidazol-1-yl)ethylideneamino]-propan-2-ol (BIPO), 1,3-bis[(4-methyl-5-imidazol-1-yl)ethylideneamino]propane (BIP), {acetatoaqua[[(4-methylimidazol-5-yl)methylene]histamine]-copper(II)} perchlorate, Schiff base condensates with tris(2-aminoethyl)amine (tren) or tris(3-aminopropyl)amine (trpn). It is also used in the synthesis of imidazolate-based metal organic framework (MOF) membrane, 3,11-bis(4-imidazolylmethyl)-7-methyl-3,7,11,17-tetraazabicyclo[11.3.1]hepta-deca-1(17),13,15-triene, a 4-imidazolyl derivative of a 14-membered tetraazamacrocycle and Cu(II) eight and six coordinate compounds.
Solubility
Soluble in water, (23 g/L) (25°C).
Notes
Air sensitive. Store away from oxidizing agents and air. Keep the container tightly closed and place it in a cool, dry and well ventilated condition.
Specifications
Specifications
| Melting Point | 166°C to 170°C |
| Quantity | 5 g |
| Sensitivity | Air Sensitive |
| Solubility Information | Soluble in water,(23g/L) (25°C). |
| Formula Weight | 110.12 |
| Percent Purity | 99% |
| Chemical Name or Material | 5-Methylimidazole-4-carboxaldehyde |
Safety and Handling
| Hazard Category | H315-H319-H335 |
| Hazard Statement | GHS H Statement H315-H319-H335 Causes skin irritation. Causes serious eye irritation. May cause respiratory irritation. |
| Precautionary Statement | P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c |
| TSCA | No |
| Recommended Storage | Ambient temperatures; Store under Argon |
RUO – Research Use Only