missing translation for 'onlineSavingsMsg'
Learn More

5-Methylhexanoic acid, 98%

Catalog No. AAA1901209
Change view
Click to view available options
Quantity:
10 g
50 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAA1901209 10 g
AAA1901218 50 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAA1901209 Supplier Thermo Scientific Chemicals Supplier No. A1901209
Get Up to $1,800 in Chemicals with Purchase
View all promotions

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 628-46-6 | C7H14O2 | 130.19 g/mol

Ion-exclusion chromatographic separations of C1-C6 aliphatic carboxylic acids on a sulfonated styrene-divinylbenzene co-polymer resin column, high sensitive conductimetric detection is achieved with 5-methylhexanoic acid as eluent.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Ion-exclusion chromatographic separations of C1-C6 aliphatic carboxylic acids on a sulfonated styrene-divinylbenzene co-polymer resin column, high sensitive conductimetric detection is achieved with 5-methylhexanoic acid as eluent.

Solubility
Slightly soluble in water.

Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Store away from strong oxidizing agents, bases.

Chemical Identifiers

CAS 628-46-6
Molecular Formula C7H14O2
Molecular Weight (g/mol) 130.19
MDL Number MFCD00036494
InChI Key MHPUGCYGQWGLJL-UHFFFAOYSA-N
Synonym isoheptanoic acid, isoenanthic acid, 5-methylcaproic acid, isoamylacetic acid, hexanoic acid, 5-methyl, isovenanthic acid, 5-methylhexylic acid, 5-methyl-hexanoic acid, 5-methyl hexanoic acid, unii-n1ak4x34c2
PubChem CID 12344
IUPAC Name 5-methylhexanoic acid
SMILES CC(C)CCCC(O)=O

Specifications

Density 0.91
Boiling Point 216°C
Flash Point 102°C (216°F)
Odor Rancid
Refractive Index 1.422
Quantity 10 g
UN Number UN3265
Solubility Information Slightly soluble in water.
Formula Weight 130.19
Percent Purity 98%
Chemical Name or Material 5-Methylhexanoic acid
Show More Show Less
Hazard Category H290-H314-H335
Hazard Statement GHS H Statement
H314-H318
Causes severe skin burns and eye damage.
Causes serious eye damage.
Precautionary Statement P234-P260-P264b-P271-P280-P301+P330+P331-P303+P361+P353-P304+P340-P305+P351+P338-P310-P363-P390-P501c
DOTInformation Transport Hazard Class: 8; Packing Group: III; Proper Shipping Name: CORROSIVE LIQUID, ACIDIC, ORGANIC, N.O.S.
RTECSNumber MO8400900
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only