missing translation for 'onlineSavingsMsg'
Learn More

5-Methylhexanoic acid, 98%, Thermo Scientific™

Supplier:  Thermo Scientific Chemicals 457410250

Quantity 25g

 View more versions of this product

Catalog No. AC457410250

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Description

Description

This Thermo Scientific brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

Specifications

Chemical Identifiers

628-46-6
130.19
MHPUGCYGQWGLJL-UHFFFAOYSA-N
12344
CC(C)CCCC(O)=O
C7H14O2
MFCD00036494
isoheptanoic acid, isoenanthic acid, 5-methylcaproic acid, isoamylacetic acid, hexanoic acid, 5-methyl, isovenanthic acid, 5-methylhexylic acid, 5-methyl-hexanoic acid, 5-methyl hexanoic acid, unii-n1ak4x34c2
5-methylhexanoic acid

Specifications

5-Methylhexanoic acid
0.9050g/mL
>110°C
97.5% min. (GC)
CH3CH(CH3)(CH2)3CO2H
25g
isoheptanoic acid, isoenanthic acid, 5-methylcaproic acid, isoamylacetic acid, hexanoic acid, 5-methyl, isovenanthic acid, 5-methylhexylic acid, 5-methyl-hexanoic acid, 5-methyl hexanoic acid, unii-n1ak4x34c2
MHPUGCYGQWGLJL-UHFFFAOYSA-N
5-methylhexanoic acid
12344
98%
628-46-6
121.0°C (30.0 mmHg)
Authentic
C7H14O2
MFCD00036494
0.905
Solubility in water: slightly. Other solubilities: soluble in organic solvents
CC(C)CCCC(O)=O
130.19
130.18
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

GHS H Statement
May be corrosive to metals.
Causes severe skin burns and eye damage.
Causes serious eye damage.

GHS P Statement
Wear protective gloves/protective clothing/eye protection/face protection.
IF SWALLOWED: Rinse mouth.
Do NOT induce vomiting.
IF ON SKIN (or hair): Take off immediately all contaminated clothing.
Rinse skin with wat

GHS Signal Word: Danger

SDS
Documents

Documents

Promotions

Promotions

Promo
Expires: 06-30-2027
Get Up to $1,800 in Chemicals with Purchase
Spend $250 or more on Thermo Fisher Scientific chemicals and get up to $1,800 to spend on additional chemicals.

Adding...