missing translation for 'onlineSavingsMsg'
Learn More

5-Methylhexanoic acid, 98%, Thermo Scientific™

Catalog No. AC457410050
Change view
Click to view available options
Quantity:
5g
25g
100g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AC457410050 5g
AC457410250 25g
AC457411000 100g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AC457410050 Supplier Thermo Scientific Chemicals Supplier No. 457410050

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

This Thermo Scientific brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

Chemical Identifiers

CAS 628-46-6
Molecular Formula C7H14O2
Molecular Weight (g/mol) 130.19
MDL Number MFCD00036494
InChI Key MHPUGCYGQWGLJL-UHFFFAOYSA-N
Synonym isoheptanoic acid, isoenanthic acid, 5-methylcaproic acid, isoamylacetic acid, hexanoic acid, 5-methyl, isovenanthic acid, 5-methylhexylic acid, 5-methyl-hexanoic acid, 5-methyl hexanoic acid, unii-n1ak4x34c2
PubChem CID 12344
IUPAC Name 5-methylhexanoic acid
SMILES CC(C)CCCC(O)=O

Specifications

Density 0.9050g/mL
Boiling Point 121.0°C (30.0 mmHg)
Flash Point >110°C
Infrared Spectrum Authentic
Assay Percent Range 97.5% min. (GC)
Linear Formula CH3CH(CH3)(CH2)3CO2H
Quantity 5g
Specific Gravity 0.905
Solubility Information Solubility in water: slightly. Other solubilities: soluble in organic solvents
Formula Weight 130.18
Percent Purity 98%
Chemical Name or Material 5-Methylhexanoic acid
Show More Show Less
Hazard Category GHS Signal Word: Danger
Hazard Statement GHS H Statement
May be corrosive to metals.
Causes severe skin burns and eye damage.
Causes serious eye damage.
Precautionary Statement GHS P Statement
Wear protective gloves/protective clothing/eye protection/face protection.
IF SWALLOWED: Rinse mouth.
Do NOT induce vomiting.
IF ON SKIN (or hair): Take off immediately all contaminated clothing.
Rinse skin with wat

RUO – Research Use Only