missing translation for 'onlineSavingsMsg'
Learn More

5-Methylfurfural, Analytical Standard, ≥98.5% (GC), MilliporeSigma™ Supelco™

Catalog No. 111019363
Change view
Click to view available options
Quantity:
1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
111019363 1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 111019363 Supplier Merck Emd Millipore Supplier No. 913071ML

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Specifications

CAS 620-02-0
Density 1.107 g/mL (at 25°C)
Physical Form Neat
Linear Formula C6H6O2
MDL Number MFCD00003232
Boiling Point 187°C
Synonym 5-Methyl-2-furaldehyde
Quantity 1 mL
Formula Weight 110.11
Percent Purity ≥98.5% (GC)
Grade Analytical Standard
Show More Show Less