missing translation for 'onlineSavingsMsg'
Learn More
Learn More
5-Methylcytosine Hydrochloride 98.0+%, TCI America™
Supplier: TCI America M02041G
Specifications
| 5-Methylcytosine Hydrochloride | |
| White | |
| MFCD00035313 | |
| 5-methylcytosine hydrochloride, 5-methylcytosine hcl, 4-amino-5-methylpyrimidin-2 1h-one hydrochloride, 4-amino-2-hydroxy-5-methylpyrimidine, unii-232tob65nx, 6-amino-5-methyl-1h-pyrimidin-2-one hydrochloride, 4-amino-5-methyl-1h-pyrimidin-2-one hydrochloride, 5-methylcytosinehydrochloride, 5-methyl cytosine hydrochloride, 5 methylcytosine hydrochloride | |
| [H+].[Cl-].CC1=C(N)NC(=O)N=C1 | |
| 161.59 | |
| 161.59 | |
| Crystalline Powder |
| 58366-64-6 | |
| C5H8ClN3O | |
| 1 g | |
| ANWMULVRPAUPJT-UHFFFAOYSA-N | |
| hydrogen 6-amino-5-methyl-1,2-dihydropyrimidin-2-one chloride | |
| 94029 | |
| ≥98.0% (HPLC,N) |
Chemical Identifiers
| 58366-64-6 | |
| 161.59 | |
| ANWMULVRPAUPJT-UHFFFAOYSA-N | |
| 94029 | |
| [H+].[Cl-].CC1=C(N)NC(=O)N=C1 |
| C5H8ClN3O | |
| MFCD00035313 | |
| 5-methylcytosine hydrochloride, 5-methylcytosine hcl, 4-amino-5-methylpyrimidin-2 1h-one hydrochloride, 4-amino-2-hydroxy-5-methylpyrimidine, unii-232tob65nx, 6-amino-5-methyl-1h-pyrimidin-2-one hydrochloride, 4-amino-5-methyl-1h-pyrimidin-2-one hydrochloride, 5-methylcytosinehydrochloride, 5-methyl cytosine hydrochloride, 5 methylcytosine hydrochloride | |
| hydrogen 6-amino-5-methyl-1,2-dihydropyrimidin-2-one chloride |
Safety and Handling
TSCA : No