accessibility menu, dialog, popup

UserName

CH3NH2NHONHCl

CAS RN 58366-64-6

IUPAC Name: hydrogen 6-amino-5-methyl-1,2-dihydropyrimidin-2-one chloride
Synonyms: hydrogen 5-methylcytosine chloride
Molecular Weight (g/mol): 161.59
Molecular Formula: C5H8ClN3O
InChi Key: ANWMULVRPAUPJT-UHFFFAOYSA-N
SMILES: [H+].[Cl-].CC1=C(N)NC(=O)N=C1
Percent Purity 
  • (3)
Physical Form 
  • (3)
Quantity 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results