missing translation for 'onlineSavingsMsg'
Learn More

5-Methylcytosine Hydrochloride 98.0+%, TCI America™

Catalog No. M02045G
Change view
Click to view available options
Quantity:
100 mg
1 g
5 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
M02045G 5 g
M0204100MG 100 mg
M02041G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. M02045G Supplier TCI America Supplier No. M02045G

Chemical Identifiers

CAS 58366-64-6
Molecular Formula C5H8ClN3O
Molecular Weight (g/mol) 161.59
MDL Number MFCD00035313
InChI Key ANWMULVRPAUPJT-UHFFFAOYSA-N
Synonym 5-methylcytosine hydrochloride, 5-methylcytosine hcl, 4-amino-5-methylpyrimidin-2 1h-one hydrochloride, 4-amino-2-hydroxy-5-methylpyrimidine, unii-232tob65nx, 6-amino-5-methyl-1h-pyrimidin-2-one hydrochloride, 4-amino-5-methyl-1h-pyrimidin-2-one hydrochloride, 5-methylcytosinehydrochloride, 5-methyl cytosine hydrochloride, 5 methylcytosine hydrochloride
PubChem CID 94029
IUPAC Name hydrogen 6-amino-5-methyl-1,2-dihydropyrimidin-2-one chloride
SMILES [H+].[Cl-].CC1=C(N)NC(=O)N=C1

Specifications

Color White
Quantity 5 g
Formula Weight 161.59
Percent Purity ≥98.0% (HPLC,N)
Physical Form Crystalline Powder
Chemical Name or Material 5-Methylcytosine Hydrochloride