missing translation for 'onlineSavingsMsg'
Learn More

5-Methylbenzo[b]thiophene 98.0+%, TCI America™

Catalog No. M29741G
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
M29741G 1 g
M29745G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. M29741G Supplier TCI America Supplier No. M29741G

Chemical Identifiers

CAS 14315-14-1
Molecular Formula C9H8S
Molecular Weight (g/mol) 148.22
MDL Number MFCD00052509
InChI Key DOHZWDWNQFZIKH-UHFFFAOYSA-N
Synonym 5-methylbenzo b thiophene, benzo b thiophene, 5-methyl, 5-methylbenzothiophene, 5-methyl-benzo b thiophene, 5-methylthianaphthene, 5-methyl-benzothiophene, benzothiophene, 5-mehyl, 5-methyl benzo b thiophene
PubChem CID 84346
IUPAC Name 5-methyl-1-benzothiophene
SMILES CC1=CC=C2SC=CC2=C1

Specifications

Melting Point 37°C
Color White
Boiling Point 67°C
Quantity 1 g
Formula Weight 148.22
Percent Purity ≥98.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material 5-Methylbenzo[b]thiophene