accessibility menu, dialog, popup

UserName

CH3S

CAS RN 14315-14-1

IUPAC Name: 5-methyl-1-benzothiophene
Synonyms: benzo[b]thiophene, 5-methyl-
Molecular Weight (g/mol): 148.22
Molecular Formula: C9H8S
InChi Key: DOHZWDWNQFZIKH-UHFFFAOYSA-N
SMILES: CC1=CC=C2SC=CC2=C1
Boiling Point 
  • (2)
  • (2)
Percent Purity 
  • (2)
  • (2)
Physical Form 
  • (2)
  • (2)
Quantity 
  • (2)
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (2)