missing translation for 'onlineSavingsMsg'
Learn More

5-Methylbenzo[b]thiophene, 97%

Catalog No. p-7020637
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAA1266603 1 g
AAA1266606 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAA1266603 Supplier Thermo Scientific Chemicals Supplier No. A1266603
Only null left

CAS: 14315-14-1 | C9H8S | 148.22 g/mol

5-Methylbenzo[b]thiophene is useful in substituted thiophene for proteomics research.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
5-Methylbenzo[b]thiophene is useful in substituted thiophene for proteomics research.

Solubility
Insoluble in water.

Notes
Store in cool dry place in tightly closed container. With good ventilation. Store away from oxidizing agent.

Chemical Identifiers

CAS 14315-14-1
Molecular Formula C9H8S
Molecular Weight (g/mol) 148.22
MDL Number MFCD00052509
InChI Key DOHZWDWNQFZIKH-UHFFFAOYSA-N
Synonym 5-methylbenzo b thiophene, benzo b thiophene, 5-methyl, 5-methylbenzothiophene, 5-methyl-benzo b thiophene, 5-methylthianaphthene, 5-methyl-benzothiophene, benzothiophene, 5-mehyl, 5-methyl benzo b thiophene
PubChem CID 84346
IUPAC Name 5-methyl-1-benzothiophene
SMILES CC1=CC=C2SC=CC2=C1

Specifications

Melting Point 32°C to 36°C
Density 1.11
Boiling Point 71°C to 73°C (0.6 mmHg)
Quantity 1 g
Beilstein 109866
Solubility Information Insoluble in water.
Formula Weight 148.23
Percent Purity 97%
Chemical Name or Material 5-Methylbenzo[b]thiophene
Hazard Category H302-H315-H319-H335
Hazard Statement GHS H Statement
H301
Toxic if swallowed.
Precautionary Statement P261-P264b-P270-P271-P280-P301+P312-P302+P352-P304+P340-P305+P351+P338-P312-P330-P332+P313-P362-P501c
DOTInformation Hazard Class: 9; Packaging Group: III
EINECSNumber 238-256-2
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only